Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,5,6-Tetrabromo-p-cresol 97.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
37721-75-8
C7H4Br4O
423.724
MFCD00060318
OMVMKSWFUQZIFD-UHFFFAOYSA-N
2,3,5,6-tetrabromo-p-cresol,tetrabromo-p-cresol,2,3,5,6-tetrabromo-4-methyl-phenol,phenol,2,3,5,6-tetrabromo-4-methyl,phenol, 2,3,5,6-tetrabromo-4-methyl,tetrabrom-p-kresol,acmc-1cswl,2,3,5,6-tetrabromo-4-methylphe,4-methyl-2,3,5,6-tetrabromophenol,#
458139
2,3,5,6-tetrabromo-4-methylphenol
CC1=C(C(=C(C(=C1Br)Br)O)Br)Br
37721-75-8
C7H4Br4O
423.724
MFCD00060318
OMVMKSWFUQZIFD-UHFFFAOYSA-N
2,3,5,6-tetrabromo-p-cresol,tetrabromo-p-cresol,2,3,5,6-tetrabromo-4-methyl-phenol,phenol,2,3,5,6-tetrabromo-4-methyl,phenol, 2,3,5,6-tetrabromo-4-methyl,tetrabrom-p-kresol,acmc-1cswl,2,3,5,6-tetrabromo-4-methylphe,4-methyl-2,3,5,6-tetrabromophenol,#
458139
2,3,5,6-tetrabromo-4-methylphenol
CC1=C(C(=C(C(=C1Br)Br)O)Br)Br
Melting Point 198°C
Color White
Quantity 25g
Formula Weight 423.72
Physical Form Crystal-Powder at 20°C
Percent Purity ≥97.0% (GC)
Chemical Name or Material 2,3,5,6-Tetrabromo-p-cresol