Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,5,6-Tetrabromohydroquinone, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass Bottle
2641-89-6
C6H2Br4O2
425.696
MFCD00013970
DTFQULSULHRJOA-UHFFFAOYSA-N
tetrabromohydroquinone,1,4-dihydroxytetrabromobenzene,tetrabromoquinol,2,3,5,6-tetrabromohydroquinone,2,3,5,6-tetrabromo-1,4-benzenediol,1,2-benzenediol,3,4,5,6-tetrabromo,1,4-benzenediol, 2,3,5,6-tetrabromo,1,2,4,5-tetrabromo-3,6-dihydroxybenzene
75840
2,3,5,6-tetrabromobenzene-1,4-diol
C1(=C(C(=C(C(=C1Br)Br)O)Br)Br)O
2641-89-6
C6H2Br4O2
425.696
MFCD00013970
DTFQULSULHRJOA-UHFFFAOYSA-N
tetrabromohydroquinone,1,4-dihydroxytetrabromobenzene,tetrabromoquinol,2,3,5,6-tetrabromohydroquinone,2,3,5,6-tetrabromo-1,4-benzenediol,1,2-benzenediol,3,4,5,6-tetrabromo,1,4-benzenediol, 2,3,5,6-tetrabromo,1,2,4,5-tetrabromo-3,6-dihydroxybenzene
75840
2,3,5,6-tetrabromobenzene-1,4-diol
C1(=C(C(=C(C(=C1Br)Br)O)Br)Br)O
Packaging Glass Bottle
Quantity 25g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 425.7
Chemical Name or Material 2,3,5,6-Tetrabromohydroquinone, 98%

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 220-142-9

TSCA : TSCA