Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,5,6-Tetrachloronitrobenzene 98.0+%, TCI America™

Catalog Number p-7136370
Click to view available options
Quantity:
25 g
500 g
117-18-0
C6HCl4NO2
260.879
MFCD00007066
XQTLDIFVVHJORV-UHFFFAOYSA-N
tecnazene, tecnazen, 2,3,5,6-tetrachloronitrobenzene, arena, folosan, fusarex, myfusan, hickstor, turbostore, easytec
8330
CHEBI:82044
1,2,4,5-tetrachloro-3-nitrobenzene
C1=C(C(=C(C(=C1Cl)Cl)[N+](=O)[O-])Cl)Cl
117-18-0
C6HCl4NO2
260.879
MFCD00007066
XQTLDIFVVHJORV-UHFFFAOYSA-N
tecnazene, tecnazen, 2,3,5,6-tetrachloronitrobenzene, arena, folosan, fusarex, myfusan, hickstor, turbostore, easytec
8330
CHEBI:82044
1,2,4,5-tetrachloro-3-nitrobenzene
C1=C(C(=C(C(=C1Cl)Cl)[N+](=O)[O-])Cl)Cl
Melting Point 100°C
Quantity 25 g
UN Number 3077
Formula Weight 260.88
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,5,6-Tetrachloronitrobenzene
RTECSNumber DC0175000
TSCA No
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit