Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,5,6-Tetrafluoro-4-(trifluoromethyl)aniline, 98%, Thermo Scientific Chemicals

Catalog Number AAB2432603
Click to view available options
Quantity:
1 g
5 g
651-83-2
C7H2F7N
233.089
MFCD00091518
FJOACTZFMHZHSC-UHFFFAOYSA-N
2,3,5,6-tetrafluoro-4-trifluoromethyl aniline, 2,3,5,6-tetrafluoro-4-aminobenzotrifluoride, 4-amino-heptafluorotoluene, 4-amino-2,3,5,6-tetrafluorobenzotrifluoride, 4-aminotetrafluorobenzotrifluoride, 4-aminoheptafluorotoluene, 2,3,5,6-tetrafluoro-4-trifluoromethyl phenylamine, benzenamine, 2,3,5,6-tetrafluoro-4-trifluoromethyl, pubchem2817, yu-wasa auxiliary
616257
2,3,5,6-tetrafluoro-4-(trifluoromethyl)aniline
C1(=C(C(=C(C(=C1F)F)N)F)F)C(F)(F)F
651-83-2
C7H2F7N
233.089
MFCD00091518
FJOACTZFMHZHSC-UHFFFAOYSA-N
2,3,5,6-tetrafluoro-4-trifluoromethyl aniline, 2,3,5,6-tetrafluoro-4-aminobenzotrifluoride, 4-amino-heptafluorotoluene, 4-amino-2,3,5,6-tetrafluorobenzotrifluoride, 4-aminotetrafluorobenzotrifluoride, 4-aminoheptafluorotoluene, 2,3,5,6-tetrafluoro-4-trifluoromethyl phenylamine, benzenamine, 2,3,5,6-tetrafluoro-4-trifluoromethyl, pubchem2817, yu-wasa auxiliary
616257
2,3,5,6-tetrafluoro-4-(trifluoromethyl)aniline
C1(=C(C(=C(C(=C1F)F)N)F)F)C(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 185°C to 187°C
Refractive Index 1.43
Quantity 1 g
UN Number UN2810
Formula Weight 233.09
Percent Purity 98%
Chemical Name or Material 2,3,5,6-Tetrafluoro-4-(trifluoromethyl)aniline
Hazard Category H227-H302+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit