Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,3,5,6-Tetrafluoroanisole, 97%

Click to view available options
Quantity:
5g
25g
100g
2324-98-3
C7H4F4O
180.102
MFCD00000311
AXCOCGJDERQVDK-UHFFFAOYSA-N
2,3,5,6-tetrafluoroanisole,2,3,5,6-tetrafluoromethoxy benzene,anisole, 2,3,5,6-tetrafluoro,2,3,5,6-tetrafluoromethoxybenzene,methyl 2,3,5,6-tetrafluorophenyl ether,benzene, 1,2,4,5-tetrafluoro-3-methoxy,1,2,4,5-tetrafluoro-3-methoxy-benzene,pubchem10114,acmc-1clho,2,3,5,6-tetrafluoro-anisole
75351
1,2,4,5-tetrafluoro-3-methoxybenzene
COC1=C(C(=CC(=C1F)F)F)F
2324-98-3
C7H4F4O
180.102
MFCD00000311
AXCOCGJDERQVDK-UHFFFAOYSA-N
2,3,5,6-tetrafluoroanisole,2,3,5,6-tetrafluoromethoxy benzene,anisole, 2,3,5,6-tetrafluoro,2,3,5,6-tetrafluoromethoxybenzene,methyl 2,3,5,6-tetrafluorophenyl ether,benzene, 1,2,4,5-tetrafluoro-3-methoxy,1,2,4,5-tetrafluoro-3-methoxy-benzene,pubchem10114,acmc-1clho,2,3,5,6-tetrafluoro-anisole
75351
1,2,4,5-tetrafluoro-3-methoxybenzene
COC1=C(C(=CC(=C1F)F)F)F
Quantity 100g
Flash Point 41°C (107°F)
Boiling Point 144°C to 145°C
UN Number UN1993
Formula Weight 180.1
Percent Purity 97%
Chemical Name or Material 2,3,5,6-Tetrafluoroanisole

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.