Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,5,6-Tetrafluorobenzoic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Plastic bottle
652-18-6
C7H2F4O2
194.085
MFCD00002408
KVLBXIOFJUWSJQ-UHFFFAOYSA-N
benzoic acid, 2,3,5,6-tetrafluoro,2,3,5,6-tetrafluorobenzoicacid,pubchem1341,acmc-1avk1,benzoic acid,2,3,5,6-tetrafluoro
567533
2,3,5,6-tetrafluorobenzoic acid
C1=C(C(=C(C(=C1F)F)C(=O)O)F)F
652-18-6
C7H2F4O2
194.085
MFCD00002408
KVLBXIOFJUWSJQ-UHFFFAOYSA-N
benzoic acid, 2,3,5,6-tetrafluoro,2,3,5,6-tetrafluorobenzoicacid,pubchem1341,acmc-1avk1,benzoic acid,2,3,5,6-tetrafluoro
567533
2,3,5,6-tetrafluorobenzoic acid
C1=C(C(=C(C(=C1F)F)C(=O)O)F)F
Packaging Plastic bottle
Infrared Spectrum Authentic
Quantity 1g
Color White
Melting Point 148°C to 152°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula HC6F4CO2H
Formula Weight 194.08
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 2,3,5,6-Tetrafluorobenzoic acid, 98%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye damage.

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

TSCA : TSCA