Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,3,5,6-Tetrafluorobenzyl alcohol, 98%

Click to view available options
Quantity:
1g
5g
25g
4084-38-2
C7H4F4O
180.102
MFCD00792428
AGWVQASYTKCTCC-UHFFFAOYSA-N
2,3,5,6-tetrafluorobenzyl alcohol,2,3,5,6-tetrafluorophenyl methanol,2,3,5,6-tetrafluorobenzylalcohol,benzenemethanol, 2,3,5,6-tetrafluoro,2,3,5,6-tetrafluorophenyl methan-1-ol,2,3,5,6-tetrafluorobenyl alcohol,pubchem15529,acmc-209jh7,2,3,5,6-tetrafluorbenzyl alcohol,benzenemethanol,2,3,5,6-tetrafluoro
2734029
(2,3,5,6-tetrafluorophenyl)methanol
C1=C(C(=C(C(=C1F)F)CO)F)F
4084-38-2
C7H4F4O
180.102
MFCD00792428
AGWVQASYTKCTCC-UHFFFAOYSA-N
2,3,5,6-tetrafluorobenzyl alcohol,2,3,5,6-tetrafluorophenyl methanol,2,3,5,6-tetrafluorobenzylalcohol,benzenemethanol, 2,3,5,6-tetrafluoro,2,3,5,6-tetrafluorophenyl methan-1-ol,2,3,5,6-tetrafluorobenyl alcohol,pubchem15529,acmc-209jh7,2,3,5,6-tetrafluorbenzyl alcohol,benzenemethanol,2,3,5,6-tetrafluoro
2734029
(2,3,5,6-tetrafluorophenyl)methanol
C1=C(C(=C(C(=C1F)F)CO)F)F
Odor Recognizable
Quantity 1g
Melting Point 34°C to 38°C
Boiling Point 220°C to 225°C
Formula Weight 180.1
Percent Purity 98%
Chemical Name or Material 2,3,5,6-Tetrafluorobenzyl alcohol

GHS H Statement
H301-H319-H317
Toxic if swallowed.
Causes serious eye irritation.
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger