Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,3,5,6-Tetrafluoropyridine, 97%

Click to view available options
Quantity:
1g
5g
2875-18-5
C5HF4N
151.064
MFCD00011734
HWIPMBCMGVXOKN-UHFFFAOYSA-N
pyridine, 2,3,5,6-tetrafluoro,2,3,5,6-tetrafluoro-pyridine,2,3,5,6-tetrafluoropyridine,pubchem9303,acmc-209h4j,ksc494q4b,hwipmbcmgvxokn-uhfffaoysa,2,3,5,6-tetrafluoro-pyridine; pyridine, 2,3,5,6-tetrafluoro
137749
2,3,5,6-tetrafluoropyridine
C1=C(C(=NC(=C1F)F)F)F
2875-18-5
C5HF4N
151.064
MFCD00011734
HWIPMBCMGVXOKN-UHFFFAOYSA-N
pyridine, 2,3,5,6-tetrafluoro,2,3,5,6-tetrafluoro-pyridine,2,3,5,6-tetrafluoropyridine,pubchem9303,acmc-209h4j,ksc494q4b,hwipmbcmgvxokn-uhfffaoysa,2,3,5,6-tetrafluoro-pyridine; pyridine, 2,3,5,6-tetrafluoro
137749
2,3,5,6-tetrafluoropyridine
C1=C(C(=NC(=C1F)F)F)F
Quantity 1g
Flash Point 30°C (86°F)
Boiling Point 102°C
UN Number UN1993
Formula Weight 151.06
Percent Purity 97%
Chemical Name or Material 2,3,5,6-Tetrafluoropyridine

GHS H Statement
H301-H226-H315-H319-H335
Toxic if swallowed.
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.