Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,5,6-Tetramethyliodobenzene, 98%, Acros Organics

Click to view available options
Quantity:
5g
25g
2100-25-6
C10H13I
260.118
MFCD00013705
MCRSYYLHVQGFNR-UHFFFAOYSA-N
2,3,5,6-tetramethyliodobenzene,iododurene,3-iododurene,benzene, 3-iodo-1,2,4,5-tetramethyl,1-iodo-2,3,5,6-tetramethylbenzene,3-iodo-1,2,4,5-tetramethyl-benzene,acmc-209fgl,maybridge1_002808,3-iodo-1,4,5-tetramethylbenzene,iodo-2,3,5,6-tetramethylbenzene
75011
3-iodo-1,2,4,5-tetramethylbenzene
CC1=CC(=C(C(=C1C)I)C)C
2100-25-6
C10H13I
260.118
MFCD00013705
MCRSYYLHVQGFNR-UHFFFAOYSA-N
2,3,5,6-tetramethyliodobenzene,iododurene,3-iododurene,benzene, 3-iodo-1,2,4,5-tetramethyl,1-iodo-2,3,5,6-tetramethylbenzene,3-iodo-1,2,4,5-tetramethyl-benzene,acmc-209fgl,maybridge1_002808,3-iodo-1,4,5-tetramethylbenzene,iodo-2,3,5,6-tetramethylbenzene
75011
3-iodo-1,2,4,5-tetramethylbenzene
CC1=CC(=C(C(=C1C)I)C)C
Melting Point 78°C to 82°C
Quantity 25g
Color White
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula (CH3)4C6HI
Formula Weight 260.11
Physical Form Fine Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 2,3,5,6-Tetramethyliodobenzene, 98%
Show More Show Less

LIGHT SENSITIVE

EINECSNumber : 218-266-3

TSCA : TSCA