Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,3,5-Collidine, 99%

Click to view available options
Quantity:
5g
25g
100g
695-98-7
C8H11N
121.183
MFCD00126840
GFYHSKONPJXCDE-UHFFFAOYSA-N
2,3,5-collidine,pyridine, 2,3,5-trimethyl,2,3,5-trimethyl pyridine,pubchem15901,acmc-1aul1,2,3,5-trimethyl-pyridin,2,3,5-trimethyl-pyridine,ksc490s0f
12759
2,3,5-trimethylpyridine
CC1=CC(=C(N=C1)C)C
695-98-7
C8H11N
121.183
MFCD00126840
GFYHSKONPJXCDE-UHFFFAOYSA-N
2,3,5-collidine,pyridine, 2,3,5-trimethyl,2,3,5-trimethyl pyridine,pubchem15901,acmc-1aul1,2,3,5-trimethyl-pyridin,2,3,5-trimethyl-pyridine,ksc490s0f
12759
2,3,5-trimethylpyridine
CC1=CC(=C(N=C1)C)C
Applications
2,3,5-Collidine is omeprazole intermediate which is the drug involved in proton pump inhibitors to block the production of stomach acid. It is used in the manufacture vitamin D3.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents. Store at room temperature.
Boiling Point 182°C to 184°C
Flash Point 74°C (165°F)
Odor Unpleasant
Quantity 100g
UN Number UN2810
Beilstein 108832
Formula Weight 121.18
Percent Purity 99%
Chemical Name or Material 2,3,5-Collidine

GHS H Statement
H331-H227-H302-H312-H315-H319-H335
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.