Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,5-tri-o-benzyl-1-o-(4-nitrobenzoyl)-d-α-arabinofuranose, 98%, Acros Organics

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
31598-79-5
C33H31NO8
569.61
MFCD07366498
VBDBDZHLJKDSSB-RASQHNTCSA-N
2r,3s,4r,5r-3,4-bis benzyloxy-5-benzyloxy methyl tetrahydrofuran-2-yl 4-nitrobenzoate,2r,3s,4r,5r-3,4-bis benzyloxy-5-benzyloxy methyl oxolan-2-yl 4-nitrobenzoate,3s,4s,2r,5r-3,4-bis phenylmethoxy-5-phenylmethoxy methyl oxolan-2-yl 4-ni trobenzoate,1-o-4-nitrobenzoyl-2,3,5-tri-o-benzyl-d-arabinofuranose,2,3,5-tri-o-benzyl-1-o-4-nitrobenzoyl-d-alpha-arabinofuranose,2r,3s,4r,5r-3,4-bis phenylmethoxy-5-phenylmethoxymethyl oxolan-2-yl 4-nitrobenzoate
7157212
[(2R,3S,4R,5R)-3,4-bis(phenylmethoxy)-5-(phenylmethoxymethyl)oxolan-2-yl] 4-nitrobenzoate
C1=CC=C(C=C1)COCC2C(C(C(O2)OC(=O)C3=CC=C(C=C3)[N+](=O)[O-])OCC4=CC=CC=C4)OCC5=CC=CC=C5
31598-79-5
C33H31NO8
569.61
MFCD07366498
VBDBDZHLJKDSSB-RASQHNTCSA-N
2r,3s,4r,5r-3,4-bis benzyloxy-5-benzyloxy methyl tetrahydrofuran-2-yl 4-nitrobenzoate,2r,3s,4r,5r-3,4-bis benzyloxy-5-benzyloxy methyl oxolan-2-yl 4-nitrobenzoate,3s,4s,2r,5r-3,4-bis phenylmethoxy-5-phenylmethoxy methyl oxolan-2-yl 4-ni trobenzoate,1-o-4-nitrobenzoyl-2,3,5-tri-o-benzyl-d-arabinofuranose,2,3,5-tri-o-benzyl-1-o-4-nitrobenzoyl-d-alpha-arabinofuranose,2r,3s,4r,5r-3,4-bis phenylmethoxy-5-phenylmethoxymethyl oxolan-2-yl 4-nitrobenzoate
7157212
[(2R,3S,4R,5R)-3,4-bis(phenylmethoxy)-5-(phenylmethoxymethyl)oxolan-2-yl] 4-nitrobenzoate
C1=CC=C(C=C1)COCC2C(C(C(O2)OC(=O)C3=CC=C(C=C3)[N+](=O)[O-])OCC4=CC=CC=C4)OCC5=CC=CC=C5
Packaging Glass bottle
Infrared Spectrum Authentic
Color Beige
Quantity 1g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 569.6
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material 2, 3, 5-Tri-O-benzyl-1-O-(4-nitrobenzoyl)-D-α-arabinofuranose
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.