Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,5-Tribromothiophene, 98%, Acros Organics

Click to view available options
Quantity:
5g
3141-24-0
C4HBr3S
320.824
MFCD00014521
SKDNDSLDRLEELJ-UHFFFAOYSA-N
thiophene, 2,3,5-tribromo,pubchem5608,2,5-tribromothiophene,acmc-209hmg,thiophene,3,5-tribromo,2,3,5-tribromo thiophene,ksc225k9l,2,3,5-tribromothiophene
76592
2,3,5-tribromothiophene
C1=C(SC(=C1Br)Br)Br
3141-24-0
C4HBr3S
320.824
MFCD00014521
SKDNDSLDRLEELJ-UHFFFAOYSA-N
thiophene, 2,3,5-tribromo,pubchem5608,2,5-tribromothiophene,acmc-209hmg,thiophene,3,5-tribromo,2,3,5-tribromo thiophene,ksc225k9l,2,3,5-tribromothiophene
76592
2,3,5-tribromothiophene
C1=C(SC(=C1Br)Br)Br
Color Beige to Brown-Yellow
Infrared Spectrum Authentic
Quantity 5g
Melting Point 25°C to 29°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 320.82
Physical Form Low Melting Crystalline Mass
Percent Purity ≥97.5%
Chemical Name or Material 2, 3, 5-Tribromothiophene, 98%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning