Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,3,5-Trichloropyridine, ≥98%

Click to view available options
Quantity:
100g
500g
1kg
16063-70-0
C5H2Cl3N
182.428
MFCD00043007
CNLIIAKAAMFCJG-UHFFFAOYSA-N
pyridine, 2,3,5-trichloro,2,3,5-trichloro pyridine,2,3,5-trichloro-pyridine,pubchem1127,g00013-watson-int,acmc-1at5w,2,3,6-trichloro-pyridine,2, 3, 5-trichloropyridine,ksc179q6d,tpc-i005
27666
2,3,5-trichloropyridine
C1=C(C=NC(=C1Cl)Cl)Cl
16063-70-0
C5H2Cl3N
182.428
MFCD00043007
CNLIIAKAAMFCJG-UHFFFAOYSA-N
pyridine, 2,3,5-trichloro,2,3,5-trichloro pyridine,2,3,5-trichloro-pyridine,pubchem1127,g00013-watson-int,acmc-1at5w,2,3,6-trichloro-pyridine,2, 3, 5-trichloropyridine,ksc179q6d,tpc-i005
27666
2,3,5-trichloropyridine
C1=C(C=NC(=C1Cl)Cl)Cl
Applications
It is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
Quantity 500g
Boiling Point 219°C (literature)
Flash Point 113°C (235°F)
Color White to Yellow
Melting Point 46°C to 50°C (literature)
Beilstein 119384
Solubility Information Slightly soluble in water.
Formula Weight 182.43
Physical Form Solid
Percent Purity ≥98%
Chemical Name or Material 2,3,5-Trichloropyridine
Show More Show Less

EINECSNumber : 407-270-2

RTECSNumber : UU0525000

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold