Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2,3,5-Trichloropyridine, Thermo Scientific™
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
Specifications
Melting Point | 46°C to 50°C |
Boiling Point | 219°C |
Flash Point | 113°C |
Packaging | Glass bottle |
Quantity | 50 g |
Formula Weight | 182.44 |
Chemical Name or Material | 2, 3, 5-Trichloropyridine |
Safety and Handling
Hazard Statement | GHS H Statement: Harmful to aquatic life with long lasting effects. |
Precautionary Statement | GHS P Statement: Avoid release to the environment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction