Alfa Aesar™ 2,3,5-Triphenyl-2H-tetrazolium chloride, 98%

Manufacturer: Alfa Aesar™ A1087009

Quantity 10g

 View more versions of this product

Catalog No. AAA1087009

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 298-96-4
    Molecular Formula C19H15ClN4
    Formula Weight 334.81
    MDL Number MFCD00011963
    Beilstein 3923356
    Merck Index 14,9744
    Synonym TTC
    Chemical Name or Material 2,3,5-Triphenyl-2H-tetrazolium chloride

    Specifications

    Formula Weight 334.81
    Assay Percent Range 98%
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 298-96-4
    Merck Index 14,9744
    Recommended Storage 2°C to 8°C
    RTECS Number XF8100000
    TSCA TSCA
    MDL Number MFCD00011963
    Synonym TTC
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Molecular Formula C19H15ClN4
    Health Hazard 1 Warning
    Melting Point ≈250°C (dec.)
    Quantity 10g
    Beilstein 3923356
    Chemical Name or Material 2,3,5-Triphenyl-2H-tetrazolium chloride
    Assay 98%

    Description

    Applications
    2,3,5-Triphenyl-2H-tetrazolium chloride is used as a redox indicator. It is also used in biochemical experiments, especially to indicate cellular respiration. It is employed in autopsy pathology to assist identification of post-mortem myocardial infarctions.

    Solubility
    Soluble in water, ethanol and acetone. Insoluble in ether.

    Notes
    Hygroscopic. Incompatible with strong oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    2°C to 8°C

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit