Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,5-Triphenyl-2H-tetrazolium Chloride 98%, Acros Organics

Click to view available options
Quantity:
5g
25g
100g
298-96-4
C19H15ClN4
334.807
PKDBCJSWQUOKDO-UHFFFAOYSA-M
2,3,5-triphenyltetrazolium chloride,tetrazolium red,uroscreen,red tetrazolium,urocheck,vitastain,tetrazolium chloride,triphenyltetrazolium chloride,2,3,5-triphenyl-2h-tetrazolium chloride,tetrzolium chloride
9283
CHEBI:78019
2,3,5-triphenyltetrazol-2-ium;chloride
C1=CC=C(C=C1)C2=NN([N+](=N2)C3=CC=CC=C3)C4=CC=CC=C4.[Cl-]
298-96-4
C19H15ClN4
334.807
PKDBCJSWQUOKDO-UHFFFAOYSA-M
2,3,5-triphenyltetrazolium chloride,tetrazolium red,uroscreen,red tetrazolium,urocheck,vitastain,tetrazolium chloride,triphenyltetrazolium chloride,2,3,5-triphenyl-2h-tetrazolium chloride,tetrzolium chloride
9283
CHEBI:78019
2,3,5-triphenyltetrazol-2-ium;chloride
C1=CC=C(C=C1)C2=NN([N+](=N2)C3=CC=CC=C3)C4=CC=CC=C4.[Cl-]
Quantity 5g
CAS Min % 97.5
CAS Max % 100.0
Formula Weight 334.8
Chemical Name or Material 2, 3, 5-Triphenyl-2H-tetrazolium chloride, 98%

Warning

EINECSNumber : 206-071-6

RTECSNumber : XF8100000

TSCA : TSCA