Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,5-Triphenyl-2H-tetrazolium Chloride, BAKER™, J.T.Baker™

Click to view available options
Quantity:
5g
25g
Packaging:
Wide Mouth Amber Glass Bottle

Chemical Identifiers

298-96-4
C19H15ClN4
334.807
PKDBCJSWQUOKDO-UHFFFAOYSA-M
9283
CHEBI:78019
2,3,5-triphenyltetrazol-2-ium;chloride
C1=CC=C(C=C1)C2=NN([N+](=N2)C3=CC=CC=C3)C4=CC=CC=C4.[Cl-]

A grade of chemicals for laboratory use

Packaging Wide Mouth Amber Glass Bottle
Formula Weight 334.81
Quantity 5g
Chemical Name or Material 2,3,5-Triphenyl-2H-tetrazolium Chloride