Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,5-Triphenyl-2H-Tetrazolium Chloride, Ultrapure, Affymetrix/USB™

Click to view available options
Quantity:
10g
25g
50g
100g
500g
Packaging:
Plastic bottle
298-96-4
C19H15ClN4
334.807
MFCD00011963
PKDBCJSWQUOKDO-UHFFFAOYSA-M
2,3,5-triphenyltetrazolium chloride,tetrazolium red,uroscreen,red tetrazolium,urocheck,vitastain,tetrazolium chloride,triphenyltetrazolium chloride,2,3,5-triphenyl-2h-tetrazolium chloride,tetrzolium chloride
9283
CHEBI:78019
2,3,5-triphenyltetrazol-2-ium;chloride
C1=CC=C(C=C1)C2=NN([N+](=N2)C3=CC=CC=C3)C4=CC=CC=C4.[Cl-]
298-96-4
C19H15ClN4
334.807
MFCD00011963
PKDBCJSWQUOKDO-UHFFFAOYSA-M
2,3,5-triphenyltetrazolium chloride,tetrazolium red,uroscreen,red tetrazolium,urocheck,vitastain,tetrazolium chloride,triphenyltetrazolium chloride,2,3,5-triphenyl-2h-tetrazolium chloride,tetrzolium chloride
9283
CHEBI:78019
2,3,5-triphenyltetrazol-2-ium;chloride
C1=CC=C(C=C1)C2=NN([N+](=N2)C3=CC=CC=C3)C4=CC=CC=C4.[Cl-]
Quantity 50g
Melting Point ~250°C (decomposition)
Packaging Plastic bottle
Formula Weight 334.81
Physical Form Powder
Chemical Name or Material 2,3,5-Triphenyl-2H-Tetrazolium Chloride