Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ 2,3,5-Triphenyltetrazolium Chloride, MP Biomedicals™

2,3,5-Triphenyltetrazolium Chloride is a redox indicator.

$37.75 - $339.00

Specifications

Storage Requirements Store at +4°C, protect from light.
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MP219989201 View Documents MP Biomedicals™
0219989201
1g Each for $37.75
Add to cart
 
MP219989210 View Documents MP Biomedicals™
0219989210
10g Each for $68.90
Add to cart
 
MP219989225 View Documents MP Biomedicals™
0219989225
25g Each for $115.80
Add to cart
 
MP219989250 View Documents MP Biomedicals™
0219989250
50g Each for $198.50
Add to cart
 
MP219989280 View Documents MP Biomedicals™
0219989280
100g Each for $339.00
Add to cart
 
Description

Description

In the TTC assay (also known as TTC test or tetrazolium test), TTC is used to differentiate between metabolically active and inactive tissues. The white compound is enzymatically reduced to red TPF (1,3,5-triphenylformazan) in living tissues due to the activity of various dehydrogenases (enzymes important in oxidation of organic compounds and thus cellular metabolism), while it remains as white TTC in areas of necrosis since these enzymes have been either denatured or degraded.

Specifications

Specifications

Store at +4°C, protect from light.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit