Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2',3',6'-Trifluoropropiophenone, 97+%

Click to view available options
Quantity:
1g
5g
243666-18-4
C9H7F3O
188.149
MFCD00061222
LEDVCRMPYBDECF-UHFFFAOYSA-N
2',3',6'-trifluoropropiophenone,1-2,3,6-trifluorophenyl propan-1-one,2,3,6-trifluoropropiophenone,acmc-1cg4s,1-propanone,1-2,3,6-trifluorophenyl
2777975
1-(2,3,6-trifluorophenyl)propan-1-one
CCC(=O)C1=C(C=CC(=C1F)F)F
243666-18-4
C9H7F3O
188.149
MFCD00061222
LEDVCRMPYBDECF-UHFFFAOYSA-N
2',3',6'-trifluoropropiophenone,1-2,3,6-trifluorophenyl propan-1-one,2,3,6-trifluoropropiophenone,acmc-1cg4s,1-propanone,1-2,3,6-trifluorophenyl
2777975
1-(2,3,6-trifluorophenyl)propan-1-one
CCC(=O)C1=C(C=CC(=C1F)F)F
Quantity 5g
Formula Weight 188.15
Percent Purity ≥97%
Chemical Name or Material 2',3',6'-Trifluoropropiophenone

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures