Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Benzofuran 99.0+%, TCI America™

Catalog Number p-7118327
Click to view available options
Quantity:
5 g
25 g
271-89-6
C8H6O
118.14
MFCD00005847
IANQTJSKSUMEQM-UHFFFAOYSA-N
benzofuran, 2,3-benzofuran, coumarone, benzo b furan, cumarone, benzofurfuran, 1-oxindene, coumaron, 1-oxidene
9223
CHEBI:35260
1-benzofuran
O1C=CC2=CC=CC=C12
271-89-6
C8H6O
118.14
MFCD00005847
IANQTJSKSUMEQM-UHFFFAOYSA-N
benzofuran, 2,3-benzofuran, coumarone, benzo b furan, cumarone, benzofurfuran, 1-oxindene, coumaron, 1-oxidene
9223
CHEBI:35260
1-benzofuran
O1C=CC2=CC=CC=C12
Boiling Point 175°C
Quantity 5 g
UN Number 1993
Formula Weight 118.14
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3-Benzofuran
RTECSNumber DF6423800
TSCA Yes
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit