Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2,3-Bis(2,6-diisopropylphenylimino)butane 98.0+%, TCI America™
Click to view available options
Quantity:
200mg
1g
Specifications
Color | Undesignated |
Melting Point | 101°C |
Quantity | 200mg |
Formula Weight | 404.64 |
Physical Form | Crystal-Powder at 20°C |
Percent Purity | ≥98.0% (HPLC) |
Chemical Name or Material | 2,3-Bis(2,6-diisopropylphenylimino)butane |
Safety and Handling
TSCA : No