Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(3-Bromophenyl)naphthalene, 98%

Click to view available options
Quantity:
250mg
1g
5g
667940-23-0
C16H11Br
283.168
MFCD16658911
FWPXWVYUNHYGPE-UHFFFAOYSA-N
2-3-bromophenyl naphthalene,naphthalene, 2-3-bromophenyl,2 3-bromophenyl naphthalene,2 3-bromophenyl naphthalene,ksc279g1j,2-3-bromophenyl naphthalene
23080773
2-(3-bromophenyl)naphthalene
C1=CC=C2C=C(C=CC2=C1)C3=CC(=CC=C3)Br
667940-23-0
C16H11Br
283.168
MFCD16658911
FWPXWVYUNHYGPE-UHFFFAOYSA-N
2-3-bromophenyl naphthalene,naphthalene, 2-3-bromophenyl,2 3-bromophenyl naphthalene,2 3-bromophenyl naphthalene,ksc279g1j,2-3-bromophenyl naphthalene
23080773
2-(3-bromophenyl)naphthalene
C1=CC=C2C=C(C=CC2=C1)C3=CC(=CC=C3)Br
Quantity 5g
Beilstein 11441102
Formula Weight 283.17
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 2-(3-Bromophenyl)naphthalene