Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(3-Bromophenyl)propan-2-ol, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
30951-66-7
C9H11BrO
215.09
ZRFMJMFYMQAUDO-UHFFFAOYSA-N
2-3-bromophenyl propan-2-ol,2-3-bromophenyl-2-propanol,acmc-209hjh,2-3-bromophenyl-propan-2-ol,2-3-bromo-phenyl-propan-2-ol,3-alpha-hydroxyisopropyl-1-bromobenzene
15072383
2-(3-bromophenyl)propan-2-ol
CC(C)(C1=CC(=CC=C1)Br)O
30951-66-7
C9H11BrO
215.09
ZRFMJMFYMQAUDO-UHFFFAOYSA-N
2-3-bromophenyl propan-2-ol,2-3-bromophenyl-2-propanol,acmc-209hjh,2-3-bromophenyl-propan-2-ol,2-3-bromo-phenyl-propan-2-ol,3-alpha-hydroxyisopropyl-1-bromobenzene
15072383
2-(3-bromophenyl)propan-2-ol
CC(C)(C1=CC(=CC=C1)Br)O
Boiling Point 100°C (3.0 Torr)
Packaging Glass Bottle
Quantity 1g
Assay Percent Range 96% min. (GC)
Formula Weight 215.09
Percent Purity 97%
Chemical Name or Material 2-(3-Bromophenyl)propan-2-ol

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning