Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Butanedione monoxime, 98%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Plastic bottle
57-71-6
C4H7NO2
101.1
FSEUPUDHEBLWJY-HWKANZROSA-N
2,3-butanedione monoxime,diacetyl monoxime,diacetylmonoxime,biacetyl monoxime,biacetyl monooxime,diacetyl monooxime,diacetylmonooxime,2-oximino-3-butanone,3-oximino-2-butanone,2,3-butanedione-2-monoxime
6409633
CHEBI:4480
(3E)-3-hydroxyiminobutan-2-one
CC(=NO)C(=O)C
57-71-6
C4H7NO2
101.1
FSEUPUDHEBLWJY-HWKANZROSA-N
2,3-butanedione monoxime,diacetyl monoxime,diacetylmonoxime,biacetyl monoxime,biacetyl monooxime,diacetyl monooxime,diacetylmonooxime,2-oximino-3-butanone,3-oximino-2-butanone,2,3-butanedione-2-monoxime
6409633
CHEBI:4480
(3E)-3-hydroxyiminobutan-2-one
CC(=NO)C(=O)C
Boiling Point 185°C to 186°C
Color White
Quantity 2.5kg
Suitability Suitable for determination of urea
Infrared Spectrum Authentic
Sulfated Ash 0.1% max.
Melting Point 74°C to 76°C
Packaging Plastic bottle
Assay Percent Range 98%
Linear Formula CH3C(=NOH)COCH3
Beilstein 01,772
Solubility Information Solubility in water: 50g/L (20°C). Other solubilities: very soluble in ethanol,ether,chloroform
Formula Weight 101.1
Physical Form Crystalline Powder
Percent Purity ≥97.5% (GC)
Chemical Name or Material 2, 3-Butanedione monoxime
Show More Show Less