Alfa Aesar™ 2-(3-Butynyloxy)tetrahydropyran, 98%

Manufacturer: Alfa Aesar™ H5338514

Quantity 25g

 View more versions of this product

Catalog No. AAH5338514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 40365-61-5
    Molecular Formula C9H14O2
    Formula Weight 154.21
    MDL Number MFCD00012352
    Chemical Name or Material 2-(3-Butynyloxy)tetrahydropyran

    Specifications

    Formula Weight 154.21
    Quantity 25g
    Flash Point 73°C (163°F)
    Molecular Formula C9H14O2
    MDL Number MFCD00012352
    CAS 40365-61-5
    Health Hazard 1 Danger
    Boiling Point 92° to 95°C (18mmHg)
    Health Hazard 2 GHS H Statement
    H301-H311-H227-H332-H315-H319-H335
    Toxic if swallowed.
    Toxic in contact with skin.
    Combustible liquid.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P305+P351+P338-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 2-(3-Butynyloxy)tetrahydropyran
    Assay 98%

    Description

    Applications
    2-(3-Butynyloxy)tetrahydropyran is used as a reagent in the synthesis of Phomoidride D, a bridged, polycyclic, natural compound that has the ability to inhibit squalene synthase and can be used as an antihypercholesterolemic agent. It is also used as a reagent to prepare Rugulactone, a natural compound that is isolated from Cryptocarya rugulosa.

    Notes
    Stable under recommended storage conditions. Store away from oxidizing agents.

    GHS H Statement
    H301-H311-H227-H332-H315-H319-H335
    Toxic if swallowed.
    Toxic in contact with skin.
    Combustible liquid.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P301+P310-P305+P351+P338-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit