Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(3-Chlorophenoxy)ethanethioamide, ≥95%, Thermo Scientific™

Catalog Number spb01327da
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
35370-95-7
C8H8ClNOS
201.668
MFCD00052479
RPAOLVIADVQKNA-UHFFFAOYSA-N
2-3-chlorophenoxy ethanethioamide, 2-3-chlorophenoxy thioacetamide, 1-amino-2-3-chlorophenoxy ethane-1-thione, maybridge1_008529, 3-chlorophenoxy thioacetamide, 2-3-chlorophenoxy ethanethioamide #, ethanethioamide,2-3-chlorophenoxy
2743553
2-(3-chlorophenoxy)ethanethioamide
C1=CC(=CC(=C1)Cl)OCC(=S)N
35370-95-7
C8H8ClNOS
201.668
MFCD00052479
RPAOLVIADVQKNA-UHFFFAOYSA-N
2-3-chlorophenoxy ethanethioamide, 2-3-chlorophenoxy thioacetamide, 1-amino-2-3-chlorophenoxy ethane-1-thione, maybridge1_008529, 3-chlorophenoxy thioacetamide, 2-3-chlorophenoxy ethanethioamide #, ethanethioamide,2-3-chlorophenoxy
2743553
2-(3-chlorophenoxy)ethanethioamide
C1=CC(=CC(=C1)Cl)OCC(=S)N
CAS Min % 95.0
Assay Percent Range 95%+
Packaging Amber Glass Bottle
Linear Formula ClC6H4OCH2CSNH2
Quantity 1 g
Formula Weight 201.68
Percent Purity 95%
Chemical Name or Material 2-(3-Chlorophenoxy)ethanethioamide
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit