Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(3-Chlorophenyl)ethylamine, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
13078-79-0
C8H10ClN
155.63
MFCD00047957
NRHVNPYOTNGECT-UHFFFAOYSA-N
2-3-chlorophenyl ethylamine,2-3-chlorophenyl ethanamine,3-chlorophenethylamine,2-3-chlorophenyl ethan-1-amine,2-3-chloro-phenyl-ethylamine,3-chlorophenylethanamine,3'-chlorophenethylamine,m-chlorophenethylamine,2-m-chlorophenyl ethylamine,benzeneethanamine, 3-chloro
139381
2-(3-chlorophenyl)ethanamine
C1=CC(=CC(=C1)Cl)CCN
13078-79-0
C8H10ClN
155.63
MFCD00047957
NRHVNPYOTNGECT-UHFFFAOYSA-N
2-3-chlorophenyl ethylamine,2-3-chlorophenyl ethanamine,3-chlorophenethylamine,2-3-chlorophenyl ethan-1-amine,2-3-chloro-phenyl-ethylamine,3-chlorophenylethanamine,3'-chlorophenethylamine,m-chlorophenethylamine,2-m-chlorophenyl ethylamine,benzeneethanamine, 3-chloro
139381
2-(3-chlorophenyl)ethanamine
C1=CC(=CC(=C1)Cl)CCN
Color Yellow
Boiling Point 111.0°C to 113.0°C (12.0 mmHg)
Specific Gravity 1.119
Flash Point >109°C
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 25g
Refractive Index 1.5480 to 1.5500
Density 1.1190g/mL
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula H2N(CH2)2C6H4Cl
Formula Weight 155.63
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 2-(3-Chlorophenyl)ethylamine, 98%
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
W

GHS Signal Word: Warning

TSCA : TSCA