Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(3-Chloropropyl)-2-(4-fluorophenyl)-1,3-dioxolane 97%, Acros Organics

Click to view available options
Quantity:
250g
Packaging:
Glass bottle
3308-94-9
C12H14ClFO2
244.69
MFCD00020909
FXFDJSQOCVDXBX-UHFFFAOYSA-N
2-3-chloropropyl-2-4-fluorophenyl-1,3-dioxolane,2-3-chloropropyl-2-p-fluorophenyl-1,3-dioxolane,pubchem2065,4'-fluoro-4-chlorobutyrophenone ethylene ketal,gamma-chloro-p-fluorobutyrophenone ethylene ketal,1,1-ethylenedioxy-1-4-fluorophenyl-4-chlorobutane,4-chloro-1-4-fluorophenyl-1,1-ethylenedioxybutane,1,3-dioxolane,2-3-chloropropyl-2-4-fluorophenyl,2-3-chloropropyl-2-4-fluorophenyl-1,3-dioxalane,2-4-fluorophenyl-2-3-chloropropyl-1,3-dioxolane
2723778
2-(3-chloropropyl)-2-(4-fluorophenyl)-1,3-dioxolane
C1COC(O1)(CCCCl)C2=CC=C(C=C2)F
3308-94-9
C12H14ClFO2
244.69
MFCD00020909
FXFDJSQOCVDXBX-UHFFFAOYSA-N
2-3-chloropropyl-2-4-fluorophenyl-1,3-dioxolane,2-3-chloropropyl-2-p-fluorophenyl-1,3-dioxolane,pubchem2065,4'-fluoro-4-chlorobutyrophenone ethylene ketal,gamma-chloro-p-fluorobutyrophenone ethylene ketal,1,1-ethylenedioxy-1-4-fluorophenyl-4-chlorobutane,4-chloro-1-4-fluorophenyl-1,1-ethylenedioxybutane,1,3-dioxolane,2-3-chloropropyl-2-4-fluorophenyl,2-3-chloropropyl-2-4-fluorophenyl-1,3-dioxalane,2-4-fluorophenyl-2-3-chloropropyl-1,3-dioxolane
2723778
2-(3-chloropropyl)-2-(4-fluorophenyl)-1,3-dioxolane
C1COC(O1)(CCCCl)C2=CC=C(C=C2)F
Quantity 250g
Color Colorless to Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.5050 to 1.5075 (20°C, 589nm)
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 19, V,1, 492
Formula Weight 244.69
Physical Form Oily Liquid
Percent Purity ≥97%
Chemical Name or Material 2-(3-Chloropropyl)-2-(4-fluorophenyl)-1, 3-dioxolane, 97%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning