Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Cyclopentenopyridine, 99+%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
533-37-9
C8H9N
119.17
KRNSYSYRLQDHDK-UHFFFAOYSA-N
2,3-cyclopentenopyridine,6,7-dihydro-5h-cyclopenta b pyridine,pyrindan,2,3-cyclopenteno pyridine,5h-1-pyrindine, 6,7-dihydro,5h,6h,7h-cyclopenta b pyridine,5h-cyclopenta b pyridine, 6,7-dihydro,pubchem15900,2,3-cyclopentapyridine
68292
6,7-dihydro-5H-cyclopenta[b]pyridine
C1CC2=C(C1)N=CC=C2
533-37-9
C8H9N
119.17
KRNSYSYRLQDHDK-UHFFFAOYSA-N
2,3-cyclopentenopyridine,6,7-dihydro-5h-cyclopenta b pyridine,pyrindan,2,3-cyclopenteno pyridine,5h-1-pyrindine, 6,7-dihydro,5h,6h,7h-cyclopenta b pyridine,5h-cyclopenta b pyridine, 6,7-dihydro,pubchem15900,2,3-cyclopentapyridine
68292
6,7-dihydro-5H-cyclopenta[b]pyridine
C1CC2=C(C1)N=CC=C2
Refractive Index 1.5428 to 1.5448
Specific Gravity 1.01
Quantity 5g
Density 1.0100g/mL
Boiling Point 108.0°C to 110.0°C (66.0 mbar)
Flash Point 67°C
Packaging Glass bottle
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99% min. (GC)
Formula Weight 119.17
Percent Purity 99+%
Chemical Name or Material 2, 3-Cyclopentenopyridine
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

EINECSNumber : 208-564-1