Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Diamino-5-bromopyridine, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
38875-53-5
C5H6BrN3
188.03
MFCD00460094
YRGMYJUKFJPNPD-UHFFFAOYSA-N
2,3-diamino-5-bromopyridine,5-bromo-2,3-diaminopyridine,5-bromo-2,3-pyridinediamine,2,3-pyridinediamine, 5-bromo,3-amino-5-bromopyridin-2-yl amine,abbypharma ap-17-5972,5-brompyridin-2,3-diamin,pubchem5383,acmc-1adjl,5-brompyridin-2,3-diamine
691156
5-bromopyridine-2,3-diamine
C1=C(C(=NC=C1Br)N)N
38875-53-5
C5H6BrN3
188.03
MFCD00460094
YRGMYJUKFJPNPD-UHFFFAOYSA-N
2,3-diamino-5-bromopyridine,5-bromo-2,3-diaminopyridine,5-bromo-2,3-pyridinediamine,2,3-pyridinediamine, 5-bromo,3-amino-5-bromopyridin-2-yl amine,abbypharma ap-17-5972,5-brompyridin-2,3-diamin,pubchem5383,acmc-1adjl,5-brompyridin-2,3-diamine
691156
5-bromopyridine-2,3-diamine
C1=C(C(=NC=C1Br)N)N
Melting Point 160°C to 165°C
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 5g
Color Brown or Purple to Yellow
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Solubility Information Solubility in water: freely soluble in hot water.,freely soluble in methanol,ethyl acetate,mdc
Formula Weight 188.03
Physical Form Powder or Needles
Percent Purity 97%
Chemical Name or Material 2, 3-Diamino-5-bromopyridine
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 254-172-9

TSCA : TSCA