Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Diamino-5-chloropyridine, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
25710-20-7
C5H6ClN3
143.58
MFCD01230986
ZRCMCGQDIYNWDX-UHFFFAOYSA-N
2,3-diamino-5-chloropyridine,5-chloro-2,3-diaminopyridine,5-chloro-2,3-pyridinediamine,5-chloro-pyridine-2,3-diamine,2,3-pyridinediamine,5-chloro,pubchem5388,acmc-1cjkt,5-chloro-2,3diaminopyridine,5-chloropyridin-2,3-diamine,2,3-di-amino-5-chloropyridine
1268254
5-chloropyridine-2,3-diamine
C1=C(C(=NC=C1Cl)N)N
25710-20-7
C5H6ClN3
143.58
MFCD01230986
ZRCMCGQDIYNWDX-UHFFFAOYSA-N
2,3-diamino-5-chloropyridine,5-chloro-2,3-diaminopyridine,5-chloro-2,3-pyridinediamine,5-chloro-pyridine-2,3-diamine,2,3-pyridinediamine,5-chloro,pubchem5388,acmc-1cjkt,5-chloro-2,3diaminopyridine,5-chloropyridin-2,3-diamine,2,3-di-amino-5-chloropyridine
1268254
5-chloropyridine-2,3-diamine
C1=C(C(=NC=C1Cl)N)N
Melting Point 164°C to 173°C
Packaging Glass bottle
Color Brown to White
Infrared Spectrum Authentic
Quantity 25g
Water 0.5% max.
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Beilstein 22,II,394
Solubility Information Solubility in water: freely soluble in hot water.,freely soluble in methanol,ethyl acetate,mdc
Formula Weight 143.58
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 2, 3-Diamino-5-chloropyridine, 97%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

TSCA : TSCA