Learn More
2,3-Diaminopyridine, 98%, Thermo Scientific Chemicals
Description
2,3-Diaminopyridine was used in the synthesis of 2,3-diaminopyridinium-4-hydroxybenzoate1. It was used as internal standard in analysis of poly(amidoamine)dendrimers having different terminal functionalities by capillary electrophoresis. It was used as reagent in synthesis of imidazopyridines and organometallic complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2,3-Diaminopyridine was used in the synthesis of 2,3-diaminopyridinium-4-hydroxybenzoate1. It was used as internal standard in analysis of poly(amidoamine)dendrimers having different terminal functionalities by capillary electrophoresis. It was used as reagent in synthesis of imidazopyridines and organometallic complexes.
Solubility
Soluble in water (100 g/L).
Notes
It is sensitive to air. Incompatible with oxidizing agents.
Specifications
Melting Point | 110°C to 114°C |
Boiling Point | 195°C (55 mmHg) |
Flash Point | 205°C (401°F) |
Quantity | 5 g |
UN Number | UN2671 |
Beilstein | 109869 |
Sensitivity | Air Sensitive |
Solubility Information | Soluble in water (100g/L). |
Formula Weight | 109.13 |
Percent Purity | 98% |
Show More |
Safety and Handling
Hazard Category | H301-H312+H332-H315-H319-H335 |
Hazard Statement | GHS H Statement H300-H311-H331-H315-H319-H335 Fatal if swallowed. Toxic in contact with skin. Toxic if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: AMINOPYRIDINES |
EINECSNumber | 207-200-9 |
RTECSNumber | US7548000 |
TSCA | No |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.