Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,3-Dibromo-1-propene, tech. 85%, stab. with copper

Manufacturer:  Alfa Aesar™ A12099-22

Quantity 100g

 View more versions of this product

Catalog No. AAA1209922

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

513-31-5
199.88
878169
2,3-Dibromo-1-propene, Stabilized with copper
C3H4Br2
MFCD00000211
14,3025
Technical Grade

Specifications

141°C
100g
C3H4Br2
MFCD00000211
14,3025
85%
513-31-5
81°C (177°F)
199.88
878169
2,3-Dibromo-1-propene, Stabilized with copper
Technical Grade

Safety and Handling

GHS H Statement
H318-H341-H335-H227-H315

GHS P Statement
P210-P201-P261-P280-P281-P305+P351+P338-P304+P340-P310-P405-P501a

Danger

RTECSNumber : UC8200000

TSCA : TSCA

Recommended Storage : 2°C to 8°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit