Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dibromopropene, 80%, tech., ACROS Organics™

Click to view available options
Quantity:
10mL
50mL
250mL
Packaging:
Glass bottle
513-31-5
C3H4Br2
199.87
MFCD00000211
YMFWYDYJHRGGPF-UHFFFAOYSA-N
2,3-dibromopropene,2,3-dibromo-1-propene,1-propene, 2,3-dibromo,2-bromoallyl bromide,2,3-dibromopropylene,1,2-dibromo-2-propene,alpha-epidibromohydrin,propene, 2,3-dibromo,unii-8dne7k7z6p,ccris 6712
10552
2,3-dibromoprop-1-ene
C=C(CBr)Br
513-31-5
C3H4Br2
199.87
MFCD00000211
YMFWYDYJHRGGPF-UHFFFAOYSA-N
2,3-dibromopropene,2,3-dibromo-1-propene,1-propene, 2,3-dibromo,2-bromoallyl bromide,2,3-dibromopropylene,1,2-dibromo-2-propene,alpha-epidibromohydrin,propene, 2,3-dibromo,unii-8dne7k7z6p,ccris 6712
10552
2,3-dibromoprop-1-ene
C=C(CBr)Br
Specific Gravity 2.13
Refractive Index 1.5370 to 1.547
Boiling Point 42°C to 44°C (17.0mmHg)
Flash Point 81°C
Density 2.1300g/mL
Packaging Glass bottle
Quantity 10mL
Infrared Spectrum Authentic
Color Yellow to Brown
CAS Min % 75.0
CAS Max % 100.0
Assay Percent Range 75% min. (GC)
Linear Formula BrCH2C(Br)=CH2
Beilstein 01, 201
Merck Index 15, 3030
Solubility Information Solubility in water: insoluble
Formula Weight 199.87
Physical Form Liquid
Percent Purity 80%
Grade Technical
Chemical Name or Material 2, 3-Dibromopropene
Show More Show Less

GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Danger

EINECSNumber : 208-155-8