Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dibromopropene, 80%, ACROS Organics™

Click to view available options
Quantity:
10mL
50mL
250mL
Packaging:
Glass bottle
513-31-5
C3H4Br2
199.87
MFCD00000211
YMFWYDYJHRGGPF-UHFFFAOYSA-N
2,3-dibromopropene,2,3-dibromo-1-propene,1-propene, 2,3-dibromo,2-bromoallyl bromide,2,3-dibromopropylene,1,2-dibromo-2-propene,alpha-epidibromohydrin,propene, 2,3-dibromo,unii-8dne7k7z6p,ccris 6712
10552
2,3-dibromoprop-1-ene
C=C(CBr)Br
513-31-5
C3H4Br2
199.87
MFCD00000211
YMFWYDYJHRGGPF-UHFFFAOYSA-N
2,3-dibromopropene,2,3-dibromo-1-propene,1-propene, 2,3-dibromo,2-bromoallyl bromide,2,3-dibromopropylene,1,2-dibromo-2-propene,alpha-epidibromohydrin,propene, 2,3-dibromo,unii-8dne7k7z6p,ccris 6712
10552
2,3-dibromoprop-1-ene
C=C(CBr)Br
Boiling Point 42°C to 44°C (17.0mmHg)
Flash Point 81°C
Refractive Index 1.5370 to 1.547
Specific Gravity 2.13
Density 2.1300g/mL
Packaging Glass bottle
Color Yellow to Brown
Quantity 250mL
Infrared Spectrum Authentic
CAS Min % 75.0
CAS Max % 100.0
Assay Percent Range 80%
Linear Formula BrCH2C(Br)=CH2
Beilstein 01, 201
Merck Index 15, 3030
Solubility Information Solubility in water: insoluble
Formula Weight 199.87
Physical Form Liquid
Percent Purity ≥75%
Grade Technical
Chemical Name or Material 2, 3-Dibromopropene
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

;GHS P StatementIF SWALLOWED: rinse mouth. Do NOT induce vomiting.IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.Wear protective gloves/protective clothing/eye protection/face protection.IF IN EYES: