Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dibromopropionyl chloride, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
18791-02-1
C3H3Br2ClO
250.32
HWKWYDXHMQQDQJ-UHFFFAOYSA-N
2,3-dibromopropionyl chloride,propanoyl chloride, 2,3-dibromo,alpha,beta-dibromopropionyl chloride,propanoyl chloride,2,3-dibromo,propionyl chloride, 2,3-dibromo,acmc-209eqo,propanoyl chloride, 2,3-dibromo-9ci,2,3-dibromopropanyl chloride,3-02-00-00572 beilstein handbook reference
86790
2,3-dibromopropanoyl chloride
C(C(C(=O)Cl)Br)Br
18791-02-1
C3H3Br2ClO
250.32
HWKWYDXHMQQDQJ-UHFFFAOYSA-N
2,3-dibromopropionyl chloride,propanoyl chloride, 2,3-dibromo,alpha,beta-dibromopropionyl chloride,propanoyl chloride,2,3-dibromo,propionyl chloride, 2,3-dibromo,acmc-209eqo,propanoyl chloride, 2,3-dibromo-9ci,2,3-dibromopropanyl chloride,3-02-00-00572 beilstein handbook reference
86790
2,3-dibromopropanoyl chloride
C(C(C(=O)Cl)Br)Br
Boiling Point 193°C
Flash Point 66°C
Specific Gravity 2.14
Density 2.1400g/mL
Color Colorless to Yellow
Quantity 25g
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula BrCH2CH(Br)COCl
Formula Weight 250.32
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 2, 3-Dibromopropionyl chloride
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
May be corrosive to metals.
Reacts violently with water.
Contact with water liberates toxic gas.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store in a dry place. Store in a closed container.
Keep only in original container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 242-575-2