Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,3-Dibromopyridine, 97%

Click to view available options
Quantity:
1g
5g
25g
13534-89-9
C5H3Br2N
236.894
MFCD00234014
SLMHHOVQRSSRCV-UHFFFAOYSA-N
pyridine, 2,3-dibromo,dibromopyridine,2,3-dibromo pyridine,pubchem5402,acmc-209c0b,2,3-dibromopyridine,ksc492s9r,tpc-py084,slmhhovqrssrcv-uhfffaoysa,abbypharma ap-12-5128
817102
2,3-dibromopyridine
C1=CC(=C(N=C1)Br)Br
13534-89-9
C5H3Br2N
236.894
MFCD00234014
SLMHHOVQRSSRCV-UHFFFAOYSA-N
pyridine, 2,3-dibromo,dibromopyridine,2,3-dibromo pyridine,pubchem5402,acmc-209c0b,2,3-dibromopyridine,ksc492s9r,tpc-py084,slmhhovqrssrcv-uhfffaoysa,abbypharma ap-12-5128
817102
2,3-dibromopyridine
C1=CC(=C(N=C1)Br)Br
Applications
It is employed as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Moisture sensitive. Store in dry condition. Keep oxidizing agents.
Quantity 1g
Boiling Point 249°C to 250°C
Melting Point 58°C to 60°C
Beilstein 109828
Solubility Information Insoluble in water.
Formula Weight 236.9
Percent Purity 97%
Chemical Name or Material 2,3-Dibromopyridine

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures