Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dichloro-5,6-dicyano-1,4-benzoquinone, 98%, Thermo Scientific Chemicals

Catalog Number ac113300100
missing translation for 'orderingAttributeHoverText'
Quantity:
10 g
100 g
500 g
Packaging:
Glass bottle
84-58-2
C8Cl2N2O2
227.01
HZNVUJQVZSTENZ-UHFFFAOYSA-N
2,3-dichloro-5,6-dicyano-1,4-benzoquinone, dichlorodicyanoquinone, dichlorodicyanobenzoquinone, dichlorodicyano-p-benzoquinone, 2,3-dichloro-5,6-dicyano-p-benzoquinone, 2,3-dichloro-5,6-dicyanobenzoquinone, 1,4-cyclohexadiene-1,2-dicarbonitrile, 4,5-dichloro-3,6-dioxo, ddq, 2,3-dichloro-5,6-dicyanoquinone, 4,5-dichloro-3,6-dioxo-1,4-cyclohexadiene-1,2-dicarbonitrile
6775
4,5-dichloro-3,6-dioxocyclohexa-1,4-diene-1,2-dicarbonitrile
C(#N)C1=C(C(=O)C(=C(C1=O)Cl)Cl)C#N
84-58-2
C8Cl2N2O2
227.01
HZNVUJQVZSTENZ-UHFFFAOYSA-N
2,3-dichloro-5,6-dicyano-1,4-benzoquinone, dichlorodicyanoquinone, dichlorodicyanobenzoquinone, dichlorodicyano-p-benzoquinone, 2,3-dichloro-5,6-dicyano-p-benzoquinone, 2,3-dichloro-5,6-dicyanobenzoquinone, 1,4-cyclohexadiene-1,2-dicarbonitrile, 4,5-dichloro-3,6-dioxo, ddq, 2,3-dichloro-5,6-dicyanoquinone, 4,5-dichloro-3,6-dioxo-1,4-cyclohexadiene-1,2-dicarbonitrile
6775
4,5-dichloro-3,6-dioxocyclohexa-1,4-diene-1,2-dicarbonitrile
C(#N)C1=C(C(=O)C(=C(C1=O)Cl)Cl)C#N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 210.0°C to 217.0°C
Color Yellow to Orange
Loss on Drying 0.5% max. (1 g, 80°C, 2 hrs, 40mm Hg vacuum)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Iodometry)
Packaging Glass bottle
Quantity 10 g
Beilstein 10,902
Fieser 01,215; 02,112; 03,83; 04,130; 05,193; 06,168; 07,96; 08,153; 09,148; 10,135; 11,166; 13,104; 14,126; 15,125; 16,120
Merck Index 15,3072
Solubility Information Solubility in water: reacts. Other solubilities: soluble in benzene,dioxane,acetic acid,ethanol,slightly soluble in chloroform,methylenechloride
Formula Weight 227.01
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2,3-Dichloro-5,6-dicyano-1,4-benzoquinone
Show More Show Less
missing translation for 'healthHazard1' GHS Signal Word: Danger
missing translation for 'healthHazard2' GHS H Statement
Toxic if swallowed.
Causes serious eye damage.
Contact with water liberates toxic gas.
missing translation for 'healthHazard3' GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective glo
EINECSNumber 201-542-2

RUO – Research Use Only

missing translation for 'provideContentCorrection'

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

missing translation for 'productTitle'