Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,3-Dichloro-5-fluoropyridine, 98%

Click to view available options
Quantity:
250mg
1g
5g
185985-40-4
C5H2Cl2FN
165.976
MFCD08669666
YIYIQENCPIISLL-UHFFFAOYSA-N
pyridine,2,3-dichloro-5-fluoro,pyridine, 2,3-dichloro-5-fluoro,abbypharma ap-31-5779,2,3-dichloro-5-fluoro-pyridine
18520008
2,3-dichloro-5-fluoropyridine
C1=C(C=NC(=C1Cl)Cl)F
185985-40-4
C5H2Cl2FN
165.976
MFCD08669666
YIYIQENCPIISLL-UHFFFAOYSA-N
pyridine,2,3-dichloro-5-fluoro,pyridine, 2,3-dichloro-5-fluoro,abbypharma ap-31-5779,2,3-dichloro-5-fluoro-pyridine
18520008
2,3-dichloro-5-fluoropyridine
C1=C(C=NC(=C1Cl)Cl)F
Applications
2,3-Dichloro-5-fluoropyridine is a reagent that is used in the synthesis of 3-Chloro-5-fluoro Clodinafop Propargyl (C368645), which is an impurity of Clodinafop Propargyl.

Solubility
Slightly soluble in chloroform, methanol and water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Quantity 5g
Beilstein 13831996
Formula Weight 165.98
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 2,3-Dichloro-5-fluoropyridine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning