Alfa Aesar™ 2,3-Dichloro-6-(trifluoromethyl)aniline, 97%

Manufacturer: Alfa Aesar™ H31767MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH31767MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1017777-92-2
    Molecular Formula C7H4Cl2F3N
    Formula Weight 230.02
    MDL Number MFCD09832277
    UN Number UN3077
    Synonym 2-Amino-3,4-dichlorobenzotrifluoride
    Chemical Name or Material 2,3-Dichloro-6-(trifluoromethyl)aniline

    Specifications

    Formula Weight 230.02
    UN Number UN3077
    Quantity 250mg
    Molecular Formula C7H4Cl2F3N
    MDL Number MFCD09832277
    CAS 1017777-92-2
    Melting Point 43° to 45°C
    Synonym 2-Amino-3,4-dichlorobenzotrifluoride
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H312-H332-H315-H319
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P280h-P273-P305+P351+P338-P501a
    Wear protective gloves/clothing.
    Avoid release to the environment.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    Chemical Name or Material 2,3-Dichloro-6-(trifluoromethyl)aniline
    Assay 97%

    GHS H Statement
    H302-H312-H332-H315-H319
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280h-P273-P305+P351+P338-P501a
    Wear protective gloves/clothing.
    Avoid release to the environment.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit