Alfa Aesar™ 2,3-Dichloroanisole, 97%

Manufacturer: Alfa Aesar™ A1225718

Quantity 50g

 View more versions of this product

Catalog No. AAA1225718

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1984-59-4
    Molecular Formula C7H6Cl2O
    Formula Weight 177.03
    MDL Number MFCD00000545
    Beilstein 1909166
    Synonym 2,3-Dichloromethoxybenzene
    Chemical Name or Material 2,3-Dichloroanisole

    Specifications

    Formula Weight 177.03
    MDL Number MFCD00000545
    CAS 1984-59-4
    Synonym 2,3-Dichloromethoxybenzene
    TSCA TSCA
    Beilstein 1909166
    Flash Point >110°C (230°F)
    Boiling Point 115° to 116°C (12mmHg)
    Melting Point 31° to 33°C
    Molecular Formula C7H6Cl2O
    Quantity 50g
    Chemical Name or Material 2,3-Dichloroanisole
    Assay 97%

    Description

    Applications
    Synthesis Material Intermediates. The product has been used to develop an analytical method for the determination of biogenic halogenated anisoles.

    Solubility
    Soluble in water (86.9 mg/L).

    Notes
    Store in cool, dry conditions in well sealed containers. Store away from strong oxidizing agents, strong acids. Keep container tightly closed.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit