Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dichloroanisole, 97%, Thermo Scientific Chemicals

Catalog Number p-7082025
Click to view available options
Quantity:
5 g
50 g
1984-59-4
C7H6Cl2O
177.02
MFCD00000545
HFEASCCDHUVYKU-UHFFFAOYSA-N
2,3-dichloroanisole, benzene, 1,2-dichloro-3-methoxy, 2,3-dichloromethoxybenzene, 2,3-dichloranisol, dichloroanisole 2,3-, 1,2-dichloro-3-methoxy-benzene, dichloroanisol, 2,3-dichloro-anisol, pubchem3680, 2,3-dichloro anisole
16126
1,2-dichloro-3-methoxybenzene
COC1=CC=CC(Cl)=C1Cl
1984-59-4
C7H6Cl2O
177.02
MFCD00000545
HFEASCCDHUVYKU-UHFFFAOYSA-N
2,3-dichloroanisole, benzene, 1,2-dichloro-3-methoxy, 2,3-dichloromethoxybenzene, 2,3-dichloranisol, dichloroanisole 2,3-, 1,2-dichloro-3-methoxy-benzene, dichloroanisol, 2,3-dichloro-anisol, pubchem3680, 2,3-dichloro anisole
16126
1,2-dichloro-3-methoxybenzene
COC1=CC=CC(Cl)=C1Cl

Synthesis Material Intermediates. The product has been used to develop an analytical method for the determination of biogenic halogenated anisoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Synthesis Material Intermediates. The product has been used to develop an analytical method for the determination of biogenic halogenated anisoles.

Solubility
Soluble in water (86.9 mg/L).

Notes
Store in cool, dry conditions in well sealed containers. Store away from strong oxidizing agents, strong acids. Keep container tightly closed.
TRUSTED_SUSTAINABILITY
Melting Point 31°C to 33°C
Boiling Point 115°C to 116°C (12 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 1909166
Solubility Information Soluble in water (86.9mg/L).
Formula Weight 177.03
Percent Purity 97%
Chemical Name or Material 2,3-Dichloroanisole
Hazard Category H302+H332-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 217-853-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit