Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,3-Dichlorobenzamide, 98%

Click to view available options
Quantity:
1g
5g
5980-24-5
C7H5Cl2NO
190.023
MFCD00173938
KZKYRHRAVGWEAV-UHFFFAOYSA-N
dichlorobenzamide,acmc-20aoh8,benzamide,2,3-dichloro,benzamide, 2,3-dichloro,2,3-bis chloranyl benzamide
2735956
2,3-dichlorobenzamide
C1=CC(=C(C(=C1)Cl)Cl)C(=O)N
5980-24-5
C7H5Cl2NO
190.023
MFCD00173938
KZKYRHRAVGWEAV-UHFFFAOYSA-N
dichlorobenzamide,acmc-20aoh8,benzamide,2,3-dichloro,benzamide, 2,3-dichloro,2,3-bis chloranyl benzamide
2735956
2,3-dichlorobenzamide
C1=CC(=C(C(=C1)Cl)Cl)C(=O)N
Quantity 5g
Melting Point 165°C to 168°C
Formula Weight 190.03
Percent Purity 98%
Chemical Name or Material 2,3-Dichlorobenzamide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures