Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dichlorobenzenesulfonyl chloride, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
82417-45-6
C6H3Cl3O2S
245.51
MFCD00051844
PQODWTNHDKDHIW-UHFFFAOYSA-N
2,3-dichlorobenzene-1-sulfonyl chloride,2,3-dichlorobenzenesulphonyl chloride,2,3-dichloro-benzenesulfonyl chloride,2,3-dichlorobenzenesulphonylchloride,2,3-dichlorophenylsulfonyl chloride,benzenesulfonyl chloride, 2,3-dichloro-9ci,2,3-dichlorobenzenesulfonylchloride,pubchem5114,buttpark 9357-48,acmc-1bl9r
2734222
2,3-dichlorobenzenesulfonyl chloride
C1=CC(=C(C(=C1)Cl)Cl)S(=O)(=O)Cl
82417-45-6
C6H3Cl3O2S
245.51
MFCD00051844
PQODWTNHDKDHIW-UHFFFAOYSA-N
2,3-dichlorobenzene-1-sulfonyl chloride,2,3-dichlorobenzenesulphonyl chloride,2,3-dichloro-benzenesulfonyl chloride,2,3-dichlorobenzenesulphonylchloride,2,3-dichlorophenylsulfonyl chloride,benzenesulfonyl chloride, 2,3-dichloro-9ci,2,3-dichlorobenzenesulfonylchloride,pubchem5114,buttpark 9357-48,acmc-1bl9r
2734222
2,3-dichlorobenzenesulfonyl chloride
C1=CC(=C(C(=C1)Cl)Cl)S(=O)(=O)Cl
Infrared Spectrum Authentic
Quantity 5g
Boiling Point 140.0°C to 142.0°C (2.0mmHg)
Melting Point 61.0°C to 65.0°C
Color Tan to White
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula Cl(Cl)C6H3SO2Cl
Formula Weight 245.51
Physical Form Powder, Crystals or Chunks
Percent Purity 97%
Chemical Name or Material 2, 3-Dichlorobenzenesulfonyl chloride
Show More Show Less

GHS P Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger