Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dichlorobenzenethiol, 99+%, Acros Organics

Click to view available options
Quantity:
5g
17231-95-7
C6H4Cl2S
179.058
MFCD00041420
QGRKONUHHGBHRB-UHFFFAOYSA-N
2,3-dichlorothiophenol,2,3-dichlorobenzene-1-thiol,dichlorothiophenol,benzenethiol, dichloro,2,3-dichloro thiophenol,pubchem6796,2,3-dichiorothiophenol,2,3-dichloro-thiophenol,acmc-1cf9p,2,3-dichlorobenzene thiol
140202
2,3-dichlorobenzenethiol
C1=CC(=C(C(=C1)Cl)Cl)S
17231-95-7
C6H4Cl2S
179.058
MFCD00041420
QGRKONUHHGBHRB-UHFFFAOYSA-N
2,3-dichlorothiophenol,2,3-dichlorobenzene-1-thiol,dichlorothiophenol,benzenethiol, dichloro,2,3-dichloro thiophenol,pubchem6796,2,3-dichiorothiophenol,2,3-dichloro-thiophenol,acmc-1cf9p,2,3-dichlorobenzene thiol
140202
2,3-dichlorobenzenethiol
C1=CC(=C(C(=C1)Cl)Cl)S
Color White to Yellow
Infrared Spectrum Authentic
Melting Point 57°C to 61°C
Quantity 5g
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Linear Formula Cl2C6H3SH
Formula Weight 179.07
Physical Form Crystals
Percent Purity ≥99.0%
Chemical Name or Material 2, 3-Dichlorobenzenethiol, 99+%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning