Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dichlorobenzylamine, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
39226-95-4
C7H7Cl2N
176.05
MFCD00047927
JHBVZGONNIVXFJ-UHFFFAOYSA-N
2,3-dichlorobenzylamine,2,3-dichlorophenyl methanamine,2,3-dichloro-benzylamine,2,3-dichlorobenzyl amine,chembl13165,1-2,3-dichlorophenyl methanamine,benzenemethanamine, 2,3-dichloro,2,3-dichlorophenyl methylamine,pubchem16713,acmc-1ae0r
587625
(2,3-dichlorophenyl)methanamine
C1=CC(=C(C(=C1)Cl)Cl)CN
39226-95-4
C7H7Cl2N
176.05
MFCD00047927
JHBVZGONNIVXFJ-UHFFFAOYSA-N
2,3-dichlorobenzylamine,2,3-dichlorophenyl methanamine,2,3-dichloro-benzylamine,2,3-dichlorobenzyl amine,chembl13165,1-2,3-dichlorophenyl methanamine,benzenemethanamine, 2,3-dichloro,2,3-dichlorophenyl methylamine,pubchem16713,acmc-1ae0r
587625
(2,3-dichlorophenyl)methanamine
C1=CC(=C(C(=C1)Cl)Cl)CN
Flash Point 105°C
Boiling Point 136°C to 140°C (18.0mmHg)
Specific Gravity 1.321
Infrared Spectrum Authentic
Refractive Index 1.5820 to 1.584
Density 1.3210g/mL
Packaging Glass bottle
Quantity 5g
Color Colorless to Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula Cl2C6H3CH2NH2
Formula Weight 176.05
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 2, 3-Dichlorobenzylamine
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger