Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dichlorophenethylamine, 99+%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
34164-43-7
C8H9Cl2N
190.07
MFCD04112741
WWRLSNWKOZHYIY-UHFFFAOYSA-N
2-2,3-dichlorophenyl ethanamine,2,3-dichlorophenethylamine,2-2,3-dichlorophenyl ethyl amine,2-2,3-dichlorophenyl ethan-1-amine,2-2,3,-dichlorophenyl ethylamine,2-2,3-dichlorophenyl ethylamine,benzeneethanamine,2,3-dichloro,2-2,3-dichloro-phenyl-ethylamine
2734758
2-(2,3-dichlorophenyl)ethanamine
C1=CC(=C(C(=C1)Cl)Cl)CCN
34164-43-7
C8H9Cl2N
190.07
MFCD04112741
WWRLSNWKOZHYIY-UHFFFAOYSA-N
2-2,3-dichlorophenyl ethanamine,2,3-dichlorophenethylamine,2-2,3-dichlorophenyl ethyl amine,2-2,3-dichlorophenyl ethan-1-amine,2-2,3,-dichlorophenyl ethylamine,2-2,3-dichlorophenyl ethylamine,benzeneethanamine,2,3-dichloro,2-2,3-dichloro-phenyl-ethylamine
2734758
2-(2,3-dichlorophenyl)ethanamine
C1=CC(=C(C(=C1)Cl)Cl)CCN
Density 1.2680g/mL
Specific Gravity 1.268
Infrared Spectrum Authentic
Packaging Glass bottle
Color Colorless to Yellow
Quantity 25g
Refractive Index 1.4690 to 1.4710 (20°C, 589nm)
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99% min. (GC)
Linear Formula Cl2C6H3CH2CH2NH2
Formula Weight 190.07
Physical Form Liquid
Percent Purity 99+%
Chemical Name or Material 2, 3-Dichlorophenethylamine
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger