Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dichlorophenylboronic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
151169-74-3
C6H5BCl2O2
190.82
MFCD01075703
TYIKXPOMOYDGCS-UHFFFAOYSA-N
2,3-dichlorophenyl boronic acid,2,3-dichlorobenzeneboronic acid,2,3-dichlorophenyl boranediol,boronic acid, 2,3-dichlorophenyl,contains varying amounts of anhydride,pubchem1810,dichlorophenylboronic acid,acmc-1c0ur,dichlorobenzene boronic acid
2734661
(2,3-dichlorophenyl)boronic acid
B(C1=C(C(=CC=C1)Cl)Cl)(O)O
151169-74-3
C6H5BCl2O2
190.82
MFCD01075703
TYIKXPOMOYDGCS-UHFFFAOYSA-N
2,3-dichlorophenyl boronic acid,2,3-dichlorobenzeneboronic acid,2,3-dichlorophenyl boranediol,boronic acid, 2,3-dichlorophenyl,contains varying amounts of anhydride,pubchem1810,dichlorophenylboronic acid,acmc-1c0ur,dichlorobenzene boronic acid
2734661
(2,3-dichlorophenyl)boronic acid
B(C1=C(C(=CC=C1)Cl)Cl)(O)O
Melting Point 242°C to 244°C
Infrared Spectrum Authentic
Quantity 1g
Color White
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 190.82
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 2, 3-Dichlorophenylboronic acid
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning

Recommended Storage : Refrigerator +4°C