Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dichlorophenylboronic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
151169-74-3
C6H5BCl2O2
190.82
MFCD01075703
TYIKXPOMOYDGCS-UHFFFAOYSA-N
2,3-dichlorophenyl boronic acid,2,3-dichlorobenzeneboronic acid,2,3-dichlorophenyl boranediol,boronic acid, 2,3-dichlorophenyl,contains varying amounts of anhydride,pubchem1810,dichlorophenylboronic acid,acmc-1c0ur,dichlorobenzene boronic acid
2734661
(2,3-dichlorophenyl)boronic acid
B(C1=C(C(=CC=C1)Cl)Cl)(O)O
151169-74-3
C6H5BCl2O2
190.82
MFCD01075703
TYIKXPOMOYDGCS-UHFFFAOYSA-N
2,3-dichlorophenyl boronic acid,2,3-dichlorobenzeneboronic acid,2,3-dichlorophenyl boranediol,boronic acid, 2,3-dichlorophenyl,contains varying amounts of anhydride,pubchem1810,dichlorophenylboronic acid,acmc-1c0ur,dichlorobenzene boronic acid
2734661
(2,3-dichlorophenyl)boronic acid
B(C1=C(C(=CC=C1)Cl)Cl)(O)O
Melting Point 242°C to 244°C
Infrared Spectrum Authentic
Quantity 25g
Color White
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 190.82
Physical Form Powder
Percent Purity ≥96.0%
Chemical Name or Material 2, 3-Dichlorophenylboronic acid
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Recommended Storage : Refrigerator +4°C